2-Ethylhexyl methacrylate ≥98%

Synonyms: Methacrylic acid 2-ethylhexyl ester

290807-1L 290807-25ML
220052-328EA 149.08 USD
220052-328 220052-329
2-Ethylhexyl methacrylate ≥98%
2-Ethylhexyl methacrylate
Formula: C₁₂H₂₂O₂
MW: 198.31 g/mol
MDL Number: MFCD00009494
CAS Number: 688-84-6
EINECS: 211-708-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR