α'-Bromo-α,α,α-trifluoro-p-xylene ≥98%

Danger

290564-5G 290564-25G 290564-1G
220051-641EA 83.57 USD
220051-641 220051-640 220051-639
α'-Bromo-α,α,α-trifluoro-p-xylene ≥98%
α'-Bromo-α,α,α-trifluoro-p-xylene
Formula: C₈H₆BrF₃
Boiling Pt: 73…75 °C (0.4 mmHg)
Melting Pt: 31.5 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000403
CAS Number: 402-49-3
EINECS: 206-947-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR