α'-Bromo-α,α,α-trifluoro-m-xylene ≥98%

Danger

290556-25G 290556-5G
220051-636EA 84.22 USD
220051-636 220051-637
α'-Bromo-α,α,α-trifluoro-m-xylene ≥98%
α'-Bromo-α,α,α-trifluoro-m-xylene
Formula: C₈H₆BrF₃
Boiling Pt: 50 °C (0.3 mmHg)
Density: 1.56 g/cm³ (20 °C)
MDL Number: MFCD00000395
CAS Number: 402-23-3
EINECS: 206-938-9
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR