(3,5-Difluorophenyl)acetic acid ≥99%

Warning

Synonyms: 3,5-Difluorophenylacetic acid

290440-5G 290440-1G
220052-055EA 66.11 USD
220052-055 220052-054
(3,5-Difluorophenyl)acetic acid ≥99%
(3,5-Difluorophenyl)acetic acid
Formula: (C₆H₃F₂)CH₂COOH
MW: 172.13 g/mol
MDL Number: MFCD00010316
CAS Number: 105184-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR