(2,5-Difluorophenyl)acetic acid ≥95%

Warning

Synonyms: 2,5-Difluorophenylacetic acid

290424-1G 290424-5G
220052-048EA 29.08 USD
220052-048 220052-049
(2,5-Difluorophenyl)acetic acid ≥95%
(2,5-Difluorophenyl)acetic acid
Formula: (C₆H₃F₂)CH₂COOH
MW: 172.13 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 122…128 °C
MDL Number: MFCD00010000
CAS Number: 85068-27-5
EINECS: 285-288-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR