3,4-Difluorophenol ≥99%

Warning

290408-25G 290408-5G
220052-044EA 152.92 USD
220052-044 220052-045
3,4-Difluorophenol ≥99%
3,4-Difluorophenol
Formula: C₆H₄F₂O
MW: 130.09 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 32 - 36 °C
MDL Number: MFCD00010315
CAS Number: 2713-33-9
UN: 2921
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR