2,5-Difluorophenol ≥95%

290394-1G 290394-5G
220052-042EA 40.17 USD
220052-042 220052-043
2,5-Difluorophenol ≥95%
2,5-Difluorophenol
Formula: C₆H₄F₂O
MW: 130.09 g/mol
Boiling Pt: 166 °C (1013 hPa)
Melting Pt: 42 °C
MDL Number: MFCD00042501
CAS Number: 2713-31-7
UN: 2921
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR