3,5-Difluorobenzyl bromide ≥98%

Danger

Synonyms: α-Bromo-3,5-difluorotoluene

290319-25G 290319-5G
220052-020EA 142.15 USD
220052-020 220052-021
3,5-Difluorobenzyl bromide ≥98%
3,5-Difluorobenzyl bromide
Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Boiling Pt: 65 °C (4.5 mmHg)
Density: 1.6 g/cm³ (20 °C)
MDL Number: MFCD00010304
CAS Number: 141776-91-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR