3,5-Difluorobenzoyl chloride ≥98%

Danger

290254-25G 290254-5G
220051-771EA 213.23 USD
220051-771 220052-012
3,5-Difluorobenzoyl chloride ≥98%
3,5-Difluorobenzoyl chloride
Formula: C₇H₃ClF₂O
MW: 176.55 g/mol
Density: 1.5 g/cm³ (20 °C)
MDL Number: MFCD00010309
CAS Number: 129714-97-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR