1-Bromo-3,5-difluorobenzene ≥98%

Danger

Synonyms: 3,5-Difluorobromobenzene

290165-10G 290165-100G
220051-368EA 84.11 USD
220051-368 220051-367
1-Bromo-3,5-difluorobenzene ≥98%
1-Bromo-3,5-difluorobenzene
Formula: C₆H₃BrF₂
MW: 192.99 g/mol
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: –27 °C
Density: 1.676 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010305
CAS Number: 461-96-1
EINECS: 416-710-2
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR