2,4-Bis(trifluoromethyl)benzyl bromide ≥95%

Synonyms: 1-(Bromomethyl)-2,4-bis(trifluoromethyl)benzene

290149-1G 290149-5G
220052-009EA 74.52 USD
220052-009 220052-010
2,4-Bis(trifluoromethyl)benzyl bromide ≥95%
2,4-Bis(trifluoromethyl)benzyl bromide
Formula: C₉H₅BrF₆
MW: 307.03 g/mol
Boiling Pt: 112 °C (0.2 mmHg)
Density: 1.637 g/cm³ (20 °C)
MDL Number: MFCD00010306
CAS Number: 140690-56-8
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR