2-Amino-4-methyl-3-nitropyridine

Warning

Synonyms: 4-Methyl-3-nitro-2-pyridylamine, 4-Methyl-3-nitropyridin-2-amine, 2-Amino-3-nitro-4-picoline

290084-5G 290084-25G
220052-000EA 163.69 USD
220052-000 220051-999
2-Amino-4-methyl-3-nitropyridine
2-Amino-4-methyl-3-nitropyridine
Formula: C₆H₇N₃O₂
MW: 153.14 g/mol
Melting Pt: 140 °C
MDL Number: MFCD00006315
CAS Number: 6635-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR