Potassium benzoate ≥99%

290009-1KG 290009-250G
220051-433EA 107.69 USD
220051-433 220051-434
Potassium benzoate ≥99%
Potassium benzoate
Formula: C₇H₅KO₂
Storage Temperature: Ambient
MDL Number: MFCD00013061
CAS Number: 582-25-2
EINECS: 209-481-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR