Methylphosphonic acid

Danger

Synonyms: Methanephosphonic acid

289868-1G 289868-25G 289868-5G
220051-409EA 50.94 USD
220051-409 220051-410 220051-411
Methylphosphonic acid
Methylphosphonic acid
Formula: CH₅O₃P
MW: 96.02 g/mol
Melting Pt: 107.5 °C
Density: 1.639 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002137
CAS Number: 993-13-5
EINECS: 213-607-2
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR