3-Amino-4-hydroxybenzoic acid ≥97%

Warning

289647-25G 289647-5G
220052-284EA 95.95 USD
220052-284 220052-285
3-Amino-4-hydroxybenzoic acid ≥97%
3-Amino-4-hydroxybenzoic acid
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 201 °C
Storage Temperature: Ambient
MDL Number: MFCD00007697
CAS Number: 1571-72-8
EINECS: 216-390-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR