4,4'-Bipyridyl

Danger

Synonyms: 4,4'-Bipyridine, 4,4'-Dipyridyl

289426-25G 289426-5G
220052-261EA 156.92 USD
220052-261 220052-262
4,4'-Bipyridyl
4,4'-Bipyridyl
Formula: C₁₀H₈N₂
MW: 156.19 g/mol
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00006416
CAS Number: 553-26-4
EINECS: 209-036-3
UN: 2811
ADR: 6.1,III
Merck Index: 13,03383

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR