Cesium fluoride 99.9% (trace metals basis)

Danger

289345-100G 289345-25G 289345-5G 289345-1KG
220052-198EA 163.69 USD
220052-198 220052-199 220052-200 102292-192
Cesium fluoride 99.9% (trace metals basis)
Cesium fluoride

Used as a base in a Suzuki cross-coupling synthesis of ortho-substituted biaryls.


  • Nucleophilic fluorination of alkynyliodonium salts to form fluorovinylic compounds
  • Nucleophilic aromatic substitution (SNAr) reactions
  • Preparation of building blocks for synthesis of fluoroallylic compounds
  • Synthesis of alcohols


Reagent for nucleophilic fluorination of primary halides and sulfonates in protic media such as tert-butyl and tert-pentyl alcohols.

Formula: CsF
MW: 151.9 g/mol
Boiling Pt: 1251 °C (1013 hPa)
Melting Pt: 680 °C
Density: 4.627 g/cm3 (20 °C)
MDL Number: MFCD00010960
CAS Number: 13400-13-0
EINECS: 236-487-3
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Appearance (Color) White
Appearance (Form) Powder or Crystals or Chunk(s) or
Powder with Chunk(s) or Crystals with Chunk(s)
X-Ray Diffraction Conforms to Structure
Gravimetric Analysis (% Fluorine) 12.0 - 13.0 %
Purity (based On Trace Metals Analysis) 99.9%
Trace Metal Analysis ≤1500.0 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR