Cesium chloride ≥99.9%

Synonyms: Cesium Chloride

289329-25G 289329-100G
220052-195EA 51.69 USD
220052-195 220052-194
Cesium chloride ≥99.9%
Cesium chloride
Formula: CsCl
MW: 168.36 g/mol
Boiling Pt: 1303 °C (1013 hPa)
Melting Pt: 646 °C
Density: 3.7808 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010955
CAS Number: 7647-17-8
EINECS: 231-600-2
Merck Index: 13,02021

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR