Erbium trichloride hexahydrate ≥99.9%

Warning

289256-25G 289256-100G
220052-180EA 158.46 USD
220052-180 220052-179
Erbium trichloride hexahydrate ≥99.9%
Erbium trichloride hexahydrate
Formula: Cl₃Er·6H₂O
MW: 381.71 g/mol
Melting Pt: 168 °C
MDL Number: MFCD00149693
CAS Number: 10025-75-9
EINECS: 233-385-0

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR