Europium(III) oxide ≥99.9%

289221-25G 289221-5G
220052-174EA 1361.54 USD
220052-174 220052-175
Europium(III) oxide ≥99.9%
Europium(III) oxide
Formula: Eu₂O₃
MW: 351.92 g/mol
Melting Pt: 2325 °C
Density: 7.42 g/cm³ (20 °C)
MDL Number: MFCD00010997
CAS Number: 1308-96-9
EINECS: 215-165-6
Merck Index: 14,03904

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR