Lanthanum(III) oxide ≥99.9% Ph. Eur., BP, USP, FCC, E252

289205-50G 289205-250G
220052-171EA 158.46 USD
220052-171 220052-170
Lanthanum(III) oxide ≥99.9% Ph. Eur., BP, USP, FCC, E252
Lanthanum(III) oxide
Formula: La₂O₃
MW: 325.81 g/mol
Boiling Pt: 4200 °C (1013 hPa)
Melting Pt: 2315 °C
Density: 6.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011071
CAS Number: 1312-81-8
EINECS: 215-200-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=290fd7a767785195d587fa622db802c7b4bc8271&vid=9ebb11fade20cc0bbd323a581869b1f0dd04cb33&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR