Neodymium trinitrate hexahydrate ≥99.9%

Warning

Synonyms: Neodymium (III) nitrate hexahydrate

289175-100G 289175-25G
220052-165EA 226.15 USD
220052-165 220052-166
Neodymium trinitrate hexahydrate ≥99.9%
Neodymium trinitrate hexahydrate
Formula: N₃NdO₉·6H₂O
MW: 438.35 g/mol
Melting Pt: ∼74 °C
Density: 2.265 g/cm³ (20 °C)
MDL Number: MFCD00149799
CAS Number: 16454-60-7
EINECS: 233-153-9
UN: 1477
ADR: 5.1,III

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR