Thulium(III) oxide ≥99.9%

Warning

289167-5G 289167-1G
220052-164EA 455.38 USD
220052-164 220052-163
Thulium(III) oxide ≥99.9%
Thulium(III) oxide
Formula: O₃Tm₂
MW: 385.87 g/mol
Melting Pt: 2410 °C
Density: 8.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011285
CAS Number: 12036-44-1
EINECS: 234-851-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR