(3-Bromophenyl)acetic acid ≥98%

Synonyms: 2-(3-Bromophenyl)acetic acid

288861-5G 288861-25G 288861-1G
220052-124EA 40.82 USD
220052-124 220052-123 220052-122
(3-Bromophenyl)acetic acid ≥98%
(3-Bromophenyl)acetic acid
Formula: (C₆H₄Br)CH₂COOH
MW: 215.05 g/mol
Boiling Pt: 308
Melting Pt: 102…103 °C
MDL Number: MFCD00004330
CAS Number: 1878-67-7
EINECS: 217-522-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR