3'-Chloroacetophenone ≥98%

Warning

Synonyms: m-Chloroacetophenone, 1-(3-Chlorophenyl)ethanone

288799-25G 288799-5G
220052-111EA 31.23 USD
220052-111 220052-112
3'-Chloroacetophenone ≥98%
3'-Chloroacetophenone
Formula: C₈H₇ClO
MW: 154.6 g/mol
Boiling Pt: 241 °C (1013 hPa)
Density: 1.184 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000593
CAS Number: 99-02-5
EINECS: 202-721-8
UN: 3416
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR