1,2-Difluoro-4-nitrobenzene ≥99%

Warning

Synonyms: 3,4-Difluoronitrobenzene

288365-10G 288365-50G
220051-976EA 58.91 USD
220051-976 220051-977
1,2-Difluoro-4-nitrobenzene ≥99%
1,2-Difluoro-4-nitrobenzene
Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 80 °C (1013 hPa)
Density: 1.43 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007198
CAS Number: 369-34-6
EINECS: 206-718-2
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR