Methyltin trichloride

Synonyms: Trichloromethylstannane

288039-5G 288039-1G
220051-944EA 175.85 USD
220051-944 220051-943
Methyltin trichloride
Methyltin trichloride
Formula: CH₃Cl₃Sn
MW: 240.1 g/mol
MDL Number: MFCD00000514
CAS Number: 993-16-8
EINECS: 213-608-8
UN: 2923
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR