Dimethyltin dichloride

Danger

Synonyms: Dichlorodimethylstannane

288012-5G 288012-25G
220051-935EA 71.31 USD
220051-935 220051-934
Dimethyltin dichloride
Dimethyltin dichloride
Formula: C₂H₆Cl₂Sn
MW: 219.69 g/mol
Boiling Pt: 124 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00000517
CAS Number: 753-73-1
EINECS: 212-039-2
UN: 3146
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR