1,5-Bis(diphenylphosphino)pentane

Synonyms: Pentane-1,3-diylbis(diphenylphosphine), Pentylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)pentane

287997-5G 287997-1G
220051-933EA 68.4 USD
220051-933 220051-932
1,5-Bis(diphenylphosphino)pentane
1,5-Bis(diphenylphosphino)pentane
Formula: C₂₉H₃₀P₂
MW: 440.5 g/mol
Boiling Pt: 230 °C (0.1 -rr)
Melting Pt: 43 - 46 °C
MDL Number: MFCD00003052
CAS Number: 27721-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR