Tris(2-methylphenyl)phosphine ≥97%

Warning

Synonyms: Tris(o-tolyl)phosphine, Tri-o-tolylphosphine

287822-1G 287822-10G
220051-888EA 40.29 USD
220051-888 220051-887
Tris(2-methylphenyl)phosphine ≥97%
Tris(2-methylphenyl)phosphine
Formula: C₂₁H₂₁P
MW: 304.37 g/mol
Boiling Pt: 187…192 °C (0.05 mmHg)
Melting Pt: 125.5 °C
MDL Number: MFCD00008514
CAS Number: 6163-58-2
EINECS: 228-193-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR