trans-1,2-Cyclobutanedicarboxylic acid ≥97.0%

Warning

28684-1G 28684-5G
220051-758EA 215.38 USD
220051-758 220051-759
trans-1,2-Cyclobutanedicarboxylic acid ≥97.0%
trans-1,2-Cyclobutanedicarboxylic acid
Formula: C₆H₈O₄
MW: 144.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00068260
CAS Number: 1124-13-6
EINECS: 214-392-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR