5-Amino-2-naphthalenesulfonic acid

Synonyms: 5-Amino-2-naphthalenesulphonic Acid, 5-Amino-β-naphthalenesulphonic acid, 1-Naphthylamine-6-sulfonic acid, 1,6-Cleve's acid, 5-Aminonaphthalene-2-sulphonic acid, 5-Aminonaphthalene-2-sulfonic acid

285072-100G 285072-5G
220051-681EA 299.38 USD
220051-681 220051-682
5-Amino-2-naphthalenesulfonic acid
5-Amino-2-naphthalenesulphonic acid
Formula: C₁₀H₉NO₃S
MW: 223.25 g/mol
MDL Number: MFCD00004030
CAS Number: 119-79-9
EINECS: 204-351-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR