Indole-3-carboxylic acid

Synonyms: 1H-Indole-3-carboxylic acid

284734-5G 284734-1G
220051-404EA 57.72 USD
220051-404 220051-403
Indole-3-carboxylic acid
Indole-3-carboxylic acid
Formula: C₉H₇NO₂
MW: 161.16 g/mol
MDL Number: MFCD00005624
CAS Number: 771-50-6
EINECS: 212-231-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR