4-Allylveratrole ≥99%

Warning

Synonyms: 4-Allylveratrole, Eugenol methyll ether, 4-Prop-2-enylveratrole, 4-Allyl-1,2-Dimethoxybenzene, 4-Allyl-1,2-dimethoxybenzene, Methyl eugenol, Eugenol methyl ether, Methyleugenol

284424-5G 284424-100G
220051-601EA 31.23 USD
220051-601 220051-600
4-Allylveratrole ≥99%
4-Allylveratrole
Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: –4 °C
Density: 1.036 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008652
CAS Number: 93-15-2
EINECS: 202-223-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR