4-(Aminomethyl)benzoic acid ≥97%

283746-25G 283746-5G 283746-100G
220051-568EA 112 USD
220051-568 220051-569 220051-567
4-(Aminomethyl)benzoic acid ≥97%
4-(Aminomethyl)benzoic acid
Formula: C₈H₉NO₂
MW: 151.16 g/mol
MDL Number: MFCD00010203
CAS Number: 56-91-7
EINECS: 200-297-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR