Nickel(II) acetylacetonate

Synonyms: Bis(2,4-pentanedionato-O,O')nickel (II), Nickel (II) bis(2,4-pentanedionato-O,O'), Nickel acetylacetonate, Nickel (II) 2,4-pentanedionate

283657-25G 283657-50G 283657-5G
220051-553EA 216.92 USD
220051-553 220051-554 220051-555
Nickel(II) acetylacetonate
Nickel(II) acetylacetonate
Formula: C₁₀H₁₄NiO₄
MW: 256.91 g/mol
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: 208 °C
Density: 1.455 g/cm³ (20 °C)
MDL Number: MFCD00000024
CAS Number: 3264-82-2
EINECS: 221-875-7
Merck Index: 12,06584

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR