3-Fluoro-4-nitrotoluene ≥99%

283363-5G 283363-25G
220051-538EA 160 USD
220051-538 220051-537
3-Fluoro-4-nitrotoluene ≥99%
3-Fluoro-4-nitrotoluene
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 249 °C (1013 hPa)
Melting Pt: 53 °C
Density: 1.438 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007053
CAS Number: 446-34-4
EINECS: 207-166-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR