Platinum(II) acetylacetonate

Warning

Synonyms: Platinum (II) bis(2,4-pentanedionato-O,O'), Platinum (II) 2,4-pentanedionate, Bis(2,4-pentanedionato-O,O')platinum (II)

282782-500MG 282782-5G
220051-477EA 169.23 USD
220051-477 220051-478
Platinum(II) acetylacetonate
Platinum(II) acetylacetonate
Formula: C₁₀H₁₄O₄Pt
MW: 393.3 g/mol
Melting Pt: 250 - 252 °C
Density: ~ 1 g/cm³ (20 °C)
MDL Number: MFCD00000028
CAS Number: 15170-57-7
EINECS: 239-223-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR