Rhodium (III) acetylacetonate

Synonyms: Tris(2,4-pentanedionato-O,O')rhodium (III), Rhodium (III) tris(2,4-pentanedionato-O,O'), Rhodium (III) 2,4-pentanedionate, Rhodium(III) acetylacetonate

282774-5G 282774-1G 282774-250MG
220051-475EA 2046.15 USD
220051-475 220051-473 220051-474
Rhodium (III) acetylacetonate
Rhodium (III) acetylacetonate
Formula: C₁₅H₂₁O₆Rh
MW: 400.23 g/mol
Melting Pt: 202…205 °C
MDL Number: MFCD00083144
CAS Number: 14284-92-5
EINECS: 244-527-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR