Ruthenium(III) acetylacetonate

282766-5G 282766-1G
220051-471EA 784.62 USD
220051-471 220051-470
Ruthenium(III) acetylacetonate
Ruthenium(III) acetylacetonate
Formula: C₁₅H₂₁O₆Ru
MW: 398.4 g/mol
MDL Number: MFCD00000030
CAS Number: 14284-93-6
EINECS: 238-193-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR