Trimethoxysilane ≥95%

282626-100G 282626-25G
220050-780EA 299.08 USD
220050-780 220050-781
Trimethoxysilane ≥95%
Trimethoxysilane
Formula: C₃H₁₀O₃Si
MW: 122.2 g/mol
Boiling Pt: 81…83 °C (1013 hPa)
Melting Pt: –114.8 °C
Density: 0.9601 g/cm³ (20 °C)
MDL Number: MFCD00008340
CAS Number: 2487-90-3
EINECS: 219-637-2
UN: 3384
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR