2,6-Difluorobenzamide ≥97%

282278-10G 282278-50G
220051-135EA 38.98 USD
220051-135 220051-136
2,6-Difluorobenzamide ≥97%
2,6-Difluorobenzamide
Formula: C₇H₅F₂NO
MW: 157.12 g/mol
Boiling Pt: 284 °C (1013 hPa)
Melting Pt: 144…145.5 °C
MDL Number: MFCD00007972
CAS Number: 18063-03-1
EINECS: 241-972-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR