5-Isopropyl-2-methylphenol ≥98%

Synonyms: Carvacrol, 5-Isopropyl-o-cresol, Isothymol, 5-Isopropyl-2-methylphenol

282197-50G 282197-10G
220051-121EA 286.46 USD
220051-121 220051-120
5-Isopropyl-2-methylphenol ≥98%
5-Isopropyl-2-methylphenol
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 0.5 °C
Density: 0.98 g/cm³ (20 °C)
Flash Pt: >100 °C
MDL Number: MFCD00002236
CAS Number: 499-75-2
EINECS: 207-889-6
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR