4-Bromobiphenyl (PBB No. 3) ≥98%

Warning

Synonyms: 4-Bromodiphenyl, PBB No. 3, 4-Bromobiphenyl

281999-10G 281999-50G
220050-664EA 30.8 USD
220050-664 220050-665
4-Bromobiphenyl (PBB No. 3) ≥98%
4-Bromobiphenyl (PBB No. 3)
Formula: C₁₂H₉Br
MW: 233.11 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 90…91 °C
Density: 1.615 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00000100
CAS Number: 92-66-0
EINECS: 202-176-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR