3-Chloro-2-methylpropene ≥98%

Danger

281972-100ML 281972-1L
220050-660EA 107.15 USD
220050-660 220050-661
3-Chloro-2-methylpropene ≥98%
3-Chloro-2-methylpropene
Formula: C₄H₇Cl
MW: 90.55 g/mol
Boiling Pt: 72 °C (1013 hPa)
Melting Pt: -80 °C
Density: 0.93 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000953
CAS Number: 563-47-3
EINECS: 209-251-2
UN: 2554
ADR: 3,II
Merck Index: 13,02167

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR