Butyl butyrate ≥98%

Danger

Synonyms: Butyric acid butyl ester

281964-250ML 281964-1L
220051-314EA 36.4 USD
220051-314 220051-313
Butyl butyrate ≥98%
Butyl butyrate
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 166 °C (1013 hPa)
Melting Pt: -91.5 °C
Density: 0.872 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009450
CAS Number: 109-21-7
EINECS: 203-656-8
UN: 3272
ADR: 3,III
Merck Index: 13,01555

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR