Potassium trifluoroacetate ≥98%

Synonyms: Trifluoroacetic acid potassium salt, Potassium 2,2,2-trifluoroacetate

281883-25G 281883-100G
220051-308EA 40.85 USD
220051-308 220051-307
Potassium trifluoroacetate ≥98%
Potassium trifluoroacetate
Formula: C₂F₃KO₂
MW: 152.11 g/mol
MDL Number: MFCD00013215
CAS Number: 2923-16-2
EINECS: 220-877-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR