1-Bromo-3-iodobenzene ≥98%

Warning

Synonyms: 3-Bromoiodobenzene

280097-25G 280097-5G
220050-643EA 94.98 USD
220050-643 220050-644
1-Bromo-3-iodobenzene ≥98%
1-Bromo-3-iodobenzene
Formula: C₆H₄BrI
MW: 282.91 g/mol
Boiling Pt: 240 °C (1013 hPa)
Melting Pt: -9.3 °C
Density: 2.21 g/cm³ (20 °C)
MDL Number: MFCD00001043
CAS Number: 591-18-4
EINECS: 209-703-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR