(3,4-Dichlorophenyl)acetic acid ≥98%

Synonyms: 3,4-Dichlorophenylacetic acid

280003-25G 280003-5G
220050-633EA 124.92 USD
220050-633 220050-634
(3,4-Dichlorophenyl)acetic acid ≥98%
(3,4-Dichlorophenyl)acetic acid
Formula: (C₆H₃Cl₂)CH₂COOH
MW: 205.04 g/mol
Boiling Pt: 314 °C (1013 hPa)
Melting Pt: 91…92 °C
MDL Number: MFCD00004333
CAS Number: 5807-30-7
EINECS: 227-368-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR