2,7-Dimethylnaphthalene

279919-250MG 279919-1G
220051-053EA 64.62 USD
220051-053 220051-052
2,7-Dimethylnaphthalene
2,7-Dimethylnaphthalene
Formula: C₁₂H₁₂
MW: 156.23 g/mol
Boiling Pt: 263 °C (1013 hPa)
Melting Pt: 94 - 95 °C
Density: 1.003 g/cm³ (20 °C)
MDL Number: MFCD00004121
CAS Number: 582-16-1
EINECS: 209-477-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR